Hexaamminecobalt trichloride

Synonyms: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride

H7891-5G H7891-10G H7891-25G
220181-978EA 98.88 USD
220181-978 220181-340 220181-341
Hexaamminecobalt trichloride
Hexaamminecobalt trichloride
Formula: H₁₈CoCl₃N₆
MW: 267.47 g/mol
Density: 1.702 g/cm³ (20 °C)
MDL Number: MFCD00036304
CAS Number: 10534-89-1
EINECS: 234-103-9
Merck Index: 12,04711

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR