HEPES sodium salt

Synonyms: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, HEPES sodium, HEPES Na

H7006-2.5KG H7006-1KG H7006-500G H7006-25KG H7006-100G H7006-25G
220182-463EA 1948.15 USD
220182-463 220182-462 220182-466 220182-465 220182-461 220182-464
HEPES sodium salt
HEPES sodium salt
Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
Melting Pt: ? 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR