n-Hexadecane ≥99%, ReagentPlus®

Danger

H6703-500ML H6703-100ML H6703-1L
220181-932EA 126.92 USD
220181-932 220181-930 220181-931
n-Hexadecane ≥99%, ReagentPlus®
n-Hexadecane
Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.77331 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR