Diethylene glycol ≥99%, ReagentPlus®

Warning

Synonyms: 2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol

H26456-1KG H26456-25G H26456-4KG
220181-730EA 40.92 USD
220181-730 220181-731 220181-732
Diethylene glycol ≥99%, ReagentPlus®
Diethylene glycol
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 123 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f023bd53ecff654f6d6d398b3ee2a1bac87d56f&vid=25d8943f9c247b4203b1e1bcba59bf77dfdd9158&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR