Gadolinium trichloride hexahydrate

Warning

Synonyms: Gadolinium trichloride hexahydrate

G7532-25G G7532-5G
220180-198EA 184.62 USD
220180-198 220180-199
Gadolinium trichloride hexahydrate
Gadolinium trichloride hexahydrate
Formula: Cl₃Gd·6H₂O
MW: 371.7 g/mol
MDL Number: MFCD00149732
CAS Number: 13450-84-5
EINECS: 233-386-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR