5-Fluorouracil

Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

F6627-1G F6627-5G F6627-10G
220177-621EA 65.09 USD
220177-621 220177-622 220177-620
5-Fluorouracil
5-Fluorouracil
Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 282 - 283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,04208
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34b30f6a165337be46a329dd264fa290162179a9&vid=0a0335aa0c192ae78b4e943cb4e6a96333d3fb37&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR