5-Amino-2-methoxypyridine

A61209-25G A61209-5G
220103-985EA 130.31 USD
220103-985 220103-986
5-Amino-2-methoxypyridine
5-Amino-2-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 28…30 °C
MDL Number: MFCD00006264
CAS Number: 6628-77-9
EINECS: 229-612-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR