4-Aminobutyric acid ≥99%

A5835-25G A5835-100G A5835-10MG A5835-10G
220103-839EA 157.54 USD
220103-839 220103-836 220103-838 220103-837
4-Aminobutyric acid ≥99%
4-Aminobutyric acid
Formula: C₄H₉NO₂
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 202 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR