Diammonium hydrogen orthophosphate, Reagent Grade

A5764-500G A5764-2.5KG A5764-100G A5764-1KG
220103-830EA 45.17 USD
220103-830 220103-829 220103-827 220103-828
Diammonium hydrogen orthophosphate, Reagent Grade
di-Ammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
Melting Pt: 185 °C
Density: 1.619 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR