Acetylthiocholine iodide

Danger

Synonyms: (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide

A5751-250MG A5751-1G A5751-25G A5751-5G
220103-810EA 45.23 USD
220103-810 220103-809 220103-811 220103-812
Acetylthiocholine iodide
Acetylthiocholine iodide
Formula: H₃CC(S)O(CH₂)₂N(CH₃)₃I
MW: 289.18 g/mol
Melting Pt: 204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011819
CAS Number: 1866-15-5
EINECS: 217-474-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9024633662ddbbc8a8efbcd594cd79dbbcbb2d6&vid=c8a063af769022ab97d866f5f7547b476c5949a7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR