O-Acetylsalicylic acid ≥99.0%

Warning

Synonyms: 2-Acetoxybenzoic acid, Acetylsalicylic acid, Aspirin, Aspirin®

A5376-100G A5376-250G A5376-500G A5376-1KG
220102-890EA 32.12 USD
220102-890 220102-892 220102-893 220102-891
O-Acetylsalicylic acid ≥99.0%
o-Acetylsalicylic acid
Formula: 2-(CH₃CO₂)C₆H₄CO₂H
MW: 180.16 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 138 - 140 °C
Density: 1.39 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00002430
CAS Number: 50-78-2
EINECS: 200-064-1
Merck Index: 13,00856
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fcb60a4ee505b596cef38216a85e034985091e0&vid=5f8d5cfeb3039783869a9320ec840b8ec12c33ea&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR