4-Aminoantipyrine, Reagent Grade

Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

A4382-25G A4382-100G A4382-10G A4382-250G A4382-50G
220103-302EA 66.23 USD
220103-302 220103-299 220103-300 220103-301 220103-303
4-Aminoantipyrine, Reagent Grade
4-Aminoantipyrine
Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR