D-(-)-Arabinose

Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

A3131-10G A3131-100G A3131-25G
220101-626EA 99.32 USD
220101-626 220101-625 220101-627
D-(-)-Arabinose
D(-)-Arabinose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00135608
CAS Number: 10323-20-3
EINECS: 233-708-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR