L(+)-Asparagine

Synonyms: L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH

A0884-100G A0884-500G A0884-10MG A0884-1KG A0884-25G
220101-575EA 142.77 USD
220101-575 220101-579 220101-576 220101-577 220101-578
L(+)-Asparagine
L(+)-Asparagine
Formula: H₂NCOCH₂CH(NH₂)CO₂H
MW: 132.12 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 234…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00064401
CAS Number: 70-47-3
EINECS: 200-735-9
Merck Index: 12,00872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR