Wood's alloy (lipowitz's alloy)

Synonyms: Wood's metal, Bismuth Lead Cadmium Tin alloy (50:25:12.5:12.5 w%)

95430-500G 95430-100G
220100-922EA 410.77 USD
220100-922 220100-921
Wood's alloy (lipowitz's alloy)
Wood's alloy (lipowitz's alloy)
Melting Pt: 75 °C
Density: 9.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00198175
CAS Number: 76093-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR