1-Butyl-3-methylimidazolium bromide

95137-5G-F 95137-50G-F
220100-728EA 55.4 USD
220100-728 220100-727
1-Butyl-3-methylimidazolium bromide
1-Butyl-3-methylimidazolium bromide
Formula: C₈H₁₅BrN₂
MW: 219.12 g/mol
MDL Number: MFCD03427611
CAS Number: 85100-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR