1,1,1-Tris(hydroxymethyl)propane

Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol

93370-1KG 93370-250G
220100-345EA 57.51 USD
220100-345 220100-346
1,1,1-Tris(hydroxymethyl)propane
1,1,1-Tris(hydroxymethyl)propane
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004694
CAS Number: 77-99-6
EINECS: 201-074-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR