1-Amino-2,2,2-trifluoroethane

Synonyms: 2,2,2-Trifluoroethylamine

91692-5ML 91692-25ML
220099-914EA 328.46 USD
220099-914 220099-913
1-Amino-2,2,2-trifluoroethane
1-Amino-2,2,2-trifluoroethane
Formula: C₂H₄F₃N
MW: 99.06 g/mol
Boiling Pt: 35.85 °C (1013 hPa)
Density: 1.2452 g/cm³ (25 °C)
MDL Number: MFCD00008132
CAS Number: 753-90-2
EINECS: 212-041-3
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR