1-Butyl-3-methylimidazoliumtetrafluoroborate

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]

91508-250G 91508-5G 91508-50G
220099-903EA 764 USD
220099-903 220099-905 220099-904
1-Butyl-3-methylimidazoliumtetrafluoroborate
1-Butyl-3-methylimidazoliumtetrafluoroborate
Formula: C₈H₁₅BF₄N₂
MW: 226.02 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: –71 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD03095449
CAS Number: 174501-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR