1-Ethyl-3-methylimidazolium bromide

Synonyms: 3-Ethyl-1-methyl-1H-imidazol-3-ium bromide

89483-5G-F 89483-50G-F
220098-583EA 53.6 USD
220098-583 220098-582
1-Ethyl-3-methylimidazolium bromide
1-Ethyl-3-methylimidazolium bromide
Formula: C₆H₁₁BrN₂
MW: 191.07 g/mol
MDL Number: MFCD03427610
CAS Number: 65039-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR