1-Ethyl-3-methylimidazolium diethylphosphate, Sigma-Aldrich®

Synonyms: 1-ethyl-3-methylimidazolium diethylphosphate

713392-1KG 713392-100G
220093-151EA 1575.38 USD
220093-151 220093-150
1-Ethyl-3-methylimidazolium diethylphosphate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium diethylphosphate
Formula: C₁₀H₂₁N₂O₄P
MW: 264.26 g/mol
MDL Number: MFCD09953486
CAS Number: 848641-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR