1-Ethyl-3-methylimidazolium dicyanamide, Sigma-Aldrich®

Synonyms: Basionics VS 03, EMIM DCA

713384-1KG 713384-100G
220093-149EA 3520 USD
220093-149 220093-148
1-Ethyl-3-methylimidazolium dicyanamide, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium dicyanamide
Formula: C₈H₁₁N₅
MW: 177.21 g/mol
MDL Number: MFCD08276373
CAS Number: 370865-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR