1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate, Sigma-Aldrich®

Synonyms: 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate

711772-100G 711772-1KG
220093-013EA 392 USD
220093-013 220093-014
1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumtrifluoromethanesulphonate
Formula: C₉H₁₅F₃N₂O₃S
MW: 288.29 g/mol
MDL Number: MFCD03427620
CAS Number: 174899-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR