1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate, Sigma-Aldrich®

Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

711756-1KG 711756-100G
220093-011EA 2890 USD
220093-011 220093-010
1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate
Formula: C₇H₁₁F₃N₂O₃S
MW: 260.24 g/mol
MDL Number: MFCD00210008
CAS Number: 145022-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR