1-Ethyl-3-methylimidazoliumtetrafluoroborate, Sigma-Aldrich®

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

711721-100G 711721-1KG
220093-006EA 321 USD
220093-006 220093-007
1-Ethyl-3-methylimidazoliumtetrafluoroborate, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumtetrafluoroborate
Formula: C₆H₁₁BF₄N₂
MW: 197.97 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼15 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD00216668
CAS Number: 143314-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR