2-Bromo-6-chlorobenzoic acid ≥97%

707368-5G 707368-1G
220092-374EA 310.77 USD
220092-374 220092-373
2-Bromo-6-chlorobenzoic acid ≥97%
2-Bromo-6-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
MDL Number: MFCD00672929
CAS Number: 93224-85-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR