6-Bromo-m-anisaldehyde ≥97%

Synonyms: 2-Bromo-5-methoxybenzaldehyde

702390-1G 702390-5G
220091-865EA 29.94 USD
220091-865 220091-866
6-Bromo-m-anisaldehyde ≥97%
6-Bromo-m-anisaldehyde
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD00463817
CAS Number: 7507-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR