Methyl (±)-lactate

Danger

Synonyms: Methyl Lactate, Methyl DL-lactate, Lactic acid methyll ester, Methyl (±)-2-hydroxypropionate

69822-25ML 69822-5ML
220091-068EA 253.08 USD
220091-068 220091-069
Methyl (±)-lactate
Methyl (±)-lactate
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 35 °C (1013 hPa)
Density: 1.118 g/cm³ (0 °C)
MDL Number: MFCD00066367
CAS Number: 547-64-8
EINECS: 208-930-0
UN: 3272
ADR: 3,III
Merck Index: 12,06167

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR