Tetrakis(triphenylphosphine)palladium(0)

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

697265-500MG 697265-2G
220090-966EA 130.46 USD
220090-966 220090-965
Tetrakis(triphenylphosphine)palladium(0)
Tetrakis(triphenylphosphine)palladium(0)
Formula: C₇₂H₆₀P₄Pd
MW: 1155.58 g/mol
Melting Pt: 103…107 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR