Anthracene ≥99%, sublimated

Warning

694959-25G 694959-5G
220090-610EA 168 USD
220090-610 220090-611
Anthracene ≥99%, sublimated
Anthracene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 111 - 112 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001240
CAS Number: 120-12-7
EINECS: 204-371-1
Merck Index: 13,00687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf973bfe5e591a235e84a414db5e3a617957ab78&vid=39774282d3251d5014028c1ba9d03866f87d7733&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR