5-Bromo-o-toluonitrile ≥97%

694827-5G 694827-1G
220090-600EA 179.85 USD
220090-600 220090-599
5-Bromo-o-toluonitrile ≥97%
5-Bromo-o-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD08061920
CAS Number: 156001-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR