2-(Trimethylsilyl)benzothiazole

691569-5G 691569-1G
220090-268EA 227.23 USD
220090-268 220090-267
2-(Trimethylsilyl)benzothiazole
2-(Trimethylsilyl)benzothiazole
Formula: C₆H₁₁NSSi
MW: 157.31 g/mol
MDL Number: MFCD00068185
CAS Number: 32137-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR