1-Decyl-3-methylimidazolium chloride

Synonyms: 1-decyl-3-methylimidazolium chloride

690597-5G 690597-50G
220090-110EA 104 USD
220090-110 220090-109
1-Decyl-3-methylimidazolium chloride
1-Decyl-3-methylimidazolium chloride
Formula: C₁₄H₂₇ClN₂
MW: 258.83 g/mol
MDL Number: MFCD03427605
CAS Number: 171058-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR