3-Bromo-2-naphthol

Synonyms: 3-Bromo-2-hydroxynaphthalene, 3-Bromonaphthalen-2-ol

690120-5G 690120-1G
220090-061EA 158.31 USD
220090-061 220090-060
3-Bromo-2-naphthol
3-Bromo-2-naphthol
Formula: C₁₀H₇BrO
MW: 223.07 g/mol
MDL Number: MFCD10000951
CAS Number: 30478-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR