Halloysite nanoclay

685445-100G 685445-500G
220089-496EA 106.71 USD
220089-496 220089-497
Halloysite nanoclay
Halloysite nanoclay
Formula: Al₂Si₂O₅(OH)₄·2H₂O
MW: 294.19 g/mol
Melting Pt: 1760 °C
Density: 0.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD11045913
CAS Number: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR