3-Fluoro-4-iodopyridine

Danger

685216-5G 685216-1G
220088-472EA 734.46 USD
220088-472 220088-471
3-Fluoro-4-iodopyridine
3-Fluoro-4-iodopyridine
Formula: C₅H₃FIN
MW: 222.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03092927
CAS Number: 22282-75-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR