(2,4,5-Trifluorophenyl)acetic acid ≥97%

Synonyms: 2,4,5-Trifluorophenylacetic acid

684155-1G 684155-5G
220089-351EA 79.26 USD
220089-351 220089-352
(2,4,5-Trifluorophenyl)acetic acid ≥97%
(2,4,5-Trifluorophenyl)acetic acid
Formula: (C₆H₂F₃)CH₂COOH
MW: 190.12 g/mol
MDL Number: MFCD00082479
CAS Number: 209995-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR