2,4,5-Trichlorobenzenesulfonyl chloride ≥95%

Synonyms: 2,4,5-Trichlorobenzene-1-sulphonyl chloride

680125-5G 680125-25G
220088-807EA 73.02 USD
220088-807 220088-806
2,4,5-Trichlorobenzenesulfonyl chloride ≥95%
2,4,5-Trichlorobenzenesulphonyl chloride
Formula: C₆H₂Cl₄O₂S
MW: 279.96 g/mol
Melting Pt: 69 °C
Density: 1.88 g/cm³ (20 °C)
MDL Number: MFCD00007428
CAS Number: 15945-07-0
EINECS: 240-079-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR