Aluminium acetylacetonate

Danger

Synonyms: Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone

674753-25G 674753-5G
220087-741EA 284.62 USD
220087-741 220087-742
Aluminium acetylacetonate
Aluminium acetylacetonate
Formula: C₁₅H₂₁AlO₆
MW: 324.31 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 192…195 °C
Density: 1.204 g/cm³ (20 °C)
MDL Number: MFCD00000013
CAS Number: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR