Silver perchlorate, anhydrous

674583-5G 674583-25G
220088-283EA 103.69 USD
220088-283 220088-282
Silver perchlorate, anhydrous
Silver perchlorate
Formula: AgClO₄
Melting Pt: 486 °C
Density: 1.688 g/cm³ (25 °C)
MDL Number: MFCD00003400
CAS Number: 7783-93-9
EINECS: 232-035-4
UN: 1481
ADR: 5.1,II
Merck Index: 12,08667

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR