ω-Bromo-3-chloroacetophenone ≥97%

Synonyms: 2-Bromo-3'-chloroacetophenone, Bromomethyl m-chlorophenyl ketone, 2-Bromo-1-(3-chlorophenyl)ethanone, 3-Chlorophenacyl bromide, α-Bromo-3-chloroacetophenone

672823-5G 672823-1G
220088-119EA 649.92 USD
220088-119 220088-118
ω-Bromo-3-chloroacetophenone ≥97%
ω-Bromo-3-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00792669
CAS Number: 41011-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR