2-Chloro-6-methylbenzothiazole

Synonyms: 2-Chloro-6-methyl-1,3-benzothiazole, 2-Chloro-6-methylbenzo[d]thiazole

667374-1G 667374-5G
220086-817EA 196.15 USD
220086-817 220086-818
2-Chloro-6-methylbenzothiazole
2-Chloro-6-methylbenzothiazole
Formula: C₈H₆ClNS
MW: 183.66 g/mol
MDL Number: MFCD04448821
CAS Number: 3507-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR