4,4-Dimethylcyclohexanone ≥97%

Synonyms: 4,4-Dimethylcyclohexan-1-one

663654-1G 663654-250MG
220086-866EA 50.29 USD
220086-866 220086-867
4,4-Dimethylcyclohexanone ≥97%
4,4-Dimethylcyclohexanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
MDL Number: MFCD00234965
CAS Number: 4255-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR