2,3,6-Trifluoro-4-(trifluoromethyl)pyridine

660620-500MG 660620-100MG
220086-589EA 228.46 USD
220086-589 220086-588
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine
Formula: C₆HF₆N
MW: 201.07 g/mol
MDL Number: MFCD03840203
CAS Number: 84940-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR