Potassium-tert-butoxide ≥99.99%, sublimated

Danger

Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide

659878-25G 659878-5G
220086-518EA 293.85 USD
220086-518 220086-519
Potassium-tert-butoxide ≥99.99%, sublimated
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Melting Pt: 256…258 °C
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR