Potassium(2-naphthalene)trifluoroborate

Synonyms: Potassium 2-Naphthalenetrifluoroborate, (2-Naphthalene)trifluoroboric acid potassium salt, Potassium trifluoro(naphthalen-2-yl)borate, Potassium trifluoro(2-naphthalenyl)borate

657018-1G 657018-5G
220084-606EA 76.25 USD
220084-606 220084-607
Potassium(2-naphthalene)trifluoroborate
Potassium(2-naphthalene)trifluoroborate
Formula: C₁₀H₇BF₃K
MW: 234.07 g/mol
MDL Number: MFCD06659941
CAS Number: 668984-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR