3-Chloro-5-fluorobenzonitrile

655171-25G 655171-5G
220085-706EA 219.69 USD
220085-706 220085-707
3-Chloro-5-fluorobenzonitrile
3-Chloro-5-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
MDL Number: MFCD03407967
CAS Number: 327056-73-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR