Bismuth(III) bromide, anhydrous ≥99.999%

Danger

654981-5G 654981-1G
220085-679EA 386.15 USD
220085-679 220085-678
Bismuth(III) bromide, anhydrous ≥99.999%
Bismuth(III) bromide
Formula: BiBr₃
MW: 448.69 g/mol
Boiling Pt: 278 °C (11 -rr)
Melting Pt: 218 °C
Density: 5.72 g/cm³ (20 °C)
MDL Number: MFCD00014195
CAS Number: 7787-58-8
EINECS: 232-121-1
UN: 3260
ADR: 8,III
Merck Index: 12,01299

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR