Potassium (2-methoxyphenyl)trifluoroborate ≥95%

Synonyms: Potassium trifluoro(2-methoxyphenyl)borate, (2-Methoxyphenyl)trifluoroboric acid potassium salt

654930-5G 654930-1G
220085-672EA 263.08 USD
220085-672 220085-671
Potassium (2-methoxyphenyl)trifluoroborate ≥95%
Potassium (2-methoxyphenyl)trifluoroborate
Formula: C₇H₇BF₃KO
MW: 214.04 g/mol
MDL Number: MFCD04115756
CAS Number: 236388-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR