5-Nitrooxindole

Synonyms: 5-Nitroindolin-2-one, 5-Nitro-2-oxindole

653683-5G 653683-1G
220085-528EA 304.69 USD
220085-528 220085-527
5-Nitrooxindole
5-Nitrooxindole
Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
MDL Number: MFCD00456999
CAS Number: 20870-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR